Search
Search

Srek1| Gene Symbol: Srek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172592.2 | NP_766180.1 | 4 - 5 | 1173 | 69 | |
| GenBank mRNA | AK033354.1 | - | 4 - 5 | 1173 | 69 | |
| AK037115.1 | - | 4 - 5 | 1149 | 69 | ||
| AK039311.1 | - | 6 - 7 | 908 | 69 | ||
| AK044086.1 | - | 3 - 4 | 477 | 69 | ||
| AK052265.1 | - | 6 - 7 | 1042 | 69 | ||
| BC063761.1 | - | 7 - 8 | 918 | 69 | ||
| BC070460.1 | - | 4 - 5 | 546 | 30548562 | 69 | |
Set Membership: |
|