Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00553933_m1
          See other TRERF1 GE Assays ›
          Gene Symbol:
          Trerf1
          Gene Name:
          transcriptional regulating factor 1
          Gene Aliases:
          9430096I18Rik, AI429294, B830015H24, RAPA, Trep-132, Trep132
          Chromosome Location:
          Chr.17: 47140942 - 47359458 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00553933_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trerf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001097623.1 NP_001091092.1 6 - 7 2077 125
          NM_172622.2 NP_766210.1 3 - 4 1914 125
          GenBank mRNA AK035176.1 - 1 - 2 1684 125
          AK046822.1 - 1 - 2 1555 125
          AK154542.1 - 5 - 6 2090 125
          AK155049.1 - 5 - 6 2191 125
          AK155404.1 - 5 - 6 2088 125
          BC059215.1 - 6 - 7 2077 6412734 125
          BC108416.1 - 1 - 2 1537 5697521 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000906.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trerf1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          nucleic acid binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700008674.mm.1 NM_001097623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008674.mm.1 NM_172622.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008674.mm.1 TC1700000906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008678.mm.1 NM_001097623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008678.mm.1 NM_172622.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008678.mm.1 TC1700000906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008669.mm.1 NM_001097623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008669.mm.1 TC1700000906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008679.mm.1 NM_001097623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008679.mm.1 NM_172622.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008679.mm.1 TC1700000906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008672.mm.1 NM_001097623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008672.mm.1 TC1700000906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008675.mm.1 NM_001097623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008675.mm.1 NM_172622.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008675.mm.1 TC1700000906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700008680.mm.1 NM_001097623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008680.mm.1 NM_172622.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008680.mm.1 TC1700000906.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 603838 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline