Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mppe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172630.2 | NP_766218.1 | 5 - 6 | 1131 | 84 | |
GenBank mRNA | AK041262.1 | - | 5 - 6 | 1132 | 84 | |
AK154265.1 | - | 5 - 6 | 985 | 84 | ||
AK172000.1 | - | 5 - 6 | 970 | 84 | ||
AK172496.1 | - | 5 - 6 | 886 | 84 | ||
BC045146.1 | - | 5 - 6 | 757 | 6306245 | 84 | |
BC086456.1 | - | 5 - 6 | 804 | 4949311 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |