Search
Search
| Gene Symbol: Pdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172665.5 | NP_766253.2 | 1 - 2 | 343 | 73 | |
| GenBank mRNA | AK046805.1 | - | 1 - 2 | 344 | 73 | |
| AK085215.1 | - | 1 - 2 | 242 | 73 | ||
| AK153649.1 | - | 1 - 2 | 296 | 73 | ||
| AK163553.1 | - | 1 - 2 | 273 | 73 | ||
| BC027196.1 | - | 1 - 2 | 266 | 4458562 | 73 | |
Set Membership: |
|