| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500003892.mm.1 |
NM_172708.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003892.mm.1 |
NM_172708.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003892.mm.1 |
TC0500000390.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003894.mm.1 |
NM_172708.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003894.mm.1 |
NM_172708.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003894.mm.1 |
TC0500000390.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003893.mm.1 |
NM_172708.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003893.mm.1 |
NM_172708.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003893.mm.1 |
TC0500000390.mm.1 |
MTA TC |