| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800005154.mm.1 |
NM_001168290.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005154.mm.1 |
NM_001168290.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005154.mm.1 |
NM_172755.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005154.mm.1 |
NM_172755.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005154.mm.1 |
TC0800000741.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800005152.mm.1 |
NM_001168290.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005152.mm.1 |
NM_001168290.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005152.mm.1 |
NM_172755.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005152.mm.1 |
NM_172755.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005152.mm.1 |
TC0800000741.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800005151.mm.1 |
NM_001168290.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005151.mm.1 |
NM_001168290.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005151.mm.1 |
NM_172755.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005151.mm.1 |
NM_172755.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005151.mm.1 |
TC0800000741.mm.1 |
MTA TC |