Search
Search

Olfml2a| Gene Symbol: Olfml2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172854.2 | NP_766442.1 | 1 - 2 | 199 | 89 | |
| GenBank mRNA | AK035313.1 | - | 1 - 2 | 200 | 89 | |
| BC137871.1 | - | 1 - 2 | 127 | 8860890 | 89 | |
Set Membership: |
|