Search
Search

Olfml2a| Gene Symbol: Olfml2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172854.2 | NP_766442.1 | 4 - 5 | 780 | 108 | |
| GenBank mRNA | AK030069.1 | - | 601 | 108 | ||
| AK035313.1 | - | 4 - 5 | 781 | 108 | ||
| AK220202.1 | - | 3 - 4 | 591 | 108 | ||
| BC137871.1 | - | 4 - 5 | 708 | 8860890 | 108 | |
Set Membership: |
|