Search
Search

Parp12| Gene Symbol: Parp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172893.3 | NP_766481.2 | 2 - 3 | 671 | 73 | |
| GenBank mRNA | AK036886.1 | - | 2 - 3 | 674 | 73 | |
| AK053205.1 | - | 2 - 3 | 570 | 73 | ||
| AK081528.1 | - | 2 - 3 | 570 | 73 | ||
| AK162200.1 | - | 2 - 3 | 242 | 73 | ||
| AK206233.1 | - | 1 - 2 | 113 | 73 | ||
| BC120733.1 | - | 2 - 3 | 504 | 40129656 | 73 | |
| BC137645.1 | - | 2 - 3 | 504 | 8860664 | 73 | |
Set Membership: |
|