Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00556758_m1
          See other ANKK1 GE Assays ›
          Gene Symbol:
          Ankk1
          Gene Name:
          ankyrin repeat and kinase domain containing 1
          Gene Aliases:
          9930020N01Rik
          Chromosome Location:
          Chr.9: 49412955 - 49427791 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00556758_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172922.3 NP_766510.1 1 - 2 250 77
          GenBank mRNA AK036872.1 - 1 - 2 251 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002251.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ankk1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          phosphorylation
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158545 AK036872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158545 NM_172922.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158545 AK036872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158545 NM_172922.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294738 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66856 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113556 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121494 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158545 AK036872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158545 NM_172922.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158545 AK036872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158545 NM_172922.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26497460 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113556 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121494 - -
          Whole Mouse Genome G4122A A_51_P158545 AK036872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158545 NM_172922.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158545 AK036872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158545 NM_172922.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113139 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113556 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121494 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08128598 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08128596 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08128587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08128592 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08128589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08128591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26497489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08128601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07994030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07991508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08000908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07772477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07976199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07991499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08106885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07832801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07785412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07785410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07591219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08128597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08128588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08128584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26497419 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10593262 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593269 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593263 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593267 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593261 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593268 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593265 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593264 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593266 - -
          Affymetrix Mouse Gene 1.1 ST Array 10593260 - -
          Affymetrix Mouse Clariom S Array PSR0900018749.mm.1 NM_172922.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018749.mm.1 NM_172922.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018749.mm.1 TC0900002251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900018751.mm.1 NM_172922.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018751.mm.1 NM_172922.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900018751.mm.1 TC0900002251.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 601292 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3850563 - -
          Illumina mouse-6 v1.0 expression beadchip 3850563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221654 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221654 - -
          mouse-6 v1.1 (Illumina) scl35891.7.4_165-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850563 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35891.7.4_165-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221654 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221654 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline