| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800011178.mm.1 |
NM_173016.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011178.mm.1 |
NM_173016.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011178.mm.1 |
TC0800001360.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800011181.mm.1 |
NM_173016.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011181.mm.1 |
NM_173016.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011181.mm.1 |
TC0800001360.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800011184.mm.1 |
NM_173016.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011184.mm.1 |
NM_173016.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011184.mm.1 |
TC0800001360.mm.1 |
MTA TC |