Search
Search

Exoc6| Gene Symbol: Exoc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175353.2 | NP_780562.2 | 20 - 21 | 2217 | 70 | |
| GenBank mRNA | AK076455.1 | - | 3 - 4 | 390 | 70 | |
| AK151524.1 | - | 20 - 21 | 2218 | 70 | ||
| AK151838.1 | - | 20 - 21 | 2218 | 70 | ||
| BC025616.1 | - | 7 - 8 | 791 | 70 | ||
| BC026859.1 | - | 20 - 21 | 2170 | 2631201 | 70 | |
Set Membership: |
|