Search
Search

Plod1| Gene Symbol: Plod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011122.3 | NP_035252.1 | 8 - 9 | 943 | 60 | |
| GenBank mRNA | AF046782.1 | - | 8 - 9 | 871 | 60 | |
| AK137480.1 | - | 8 - 9 | 893 | 60 | ||
| AK157729.1 | - | 8 - 9 | 893 | 60 | ||
| AK160822.1 | - | 8 - 9 | 910 | 60 | ||
| BC006599.1 | - | 8 - 9 | 868 | 3965301 | 60 | |
Set Membership: |
|