Search
Search

Osbpl5| Gene Symbol: Osbpl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199227.1 | NP_001186156.1 | 20 - 21 | 2494 | 67 | |
| NM_024289.2 | NP_077251.1 | 20 - 21 | 2694 | 67 | ||
| GenBank mRNA | AB074008.1 | - | 20 - 21 | 2487 | 67 | |
| AJ278263.2 | - | 20 - 21 | 2694 | 67 | ||
| AK129385.1 | - | 11 - 12 | 1445 | 67 | ||
| BC079872.1 | - | 20 - 21 | 2475 | 6832073 | 67 | |
Set Membership: |
|