Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00600378_m1
          See other TPM1 GE Assays ›
          Gene Symbol:
          Tpm1
          Gene Name:
          tropomyosin 1, alpha
          Gene Aliases:
          AA986836, AI854628, TM2, Tm3, Tmpa, Tpm-1, alpha-TM
          Chromosome Location:
          Chr.9: 67022590 - 67049401 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00600378_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tpm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164248.1 NP_001157720.1 1 - 2 195 55
          NM_001164250.1 NP_001157722.1 1 - 2 195 55
          NM_001164251.1 NP_001157723.1 1 - 2 195 55
          NM_001164255.1 NP_001157727.1 1 - 2 195 55
          NM_024427.4 NP_077745.2 1 - 2 195 55
          GenBank mRNA AK002271.1 - 1 - 2 196 55
          AK003175.1 - 1 - 2 196 55
          AK032942.1 - 1 - 2 244 55
          AK077713.1 - 1 - 2 196 55
          AK168640.1 - 1 - 2 192 55
          AK168783.1 - 1 - 2 192 55
          AK169188.1 - 1 - 2 196 55
          BC026720.1 - 1 - 2 169 55
          BC132035.1 - 1 - 2 154 40130312 55
          BC132037.1 - 1 - 2 154 40130314 55
          M22479.1 - 1 - 2 168 55
          X64831.1 - 1 - 2 150 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002541.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tpm1

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          positive regulation of heart rate by epinephrine
          embryo development
          muscle filament sliding
          negative regulation of cell migration
          ruffle organization
          positive regulation of ATPase activity
          wound healing
          regulation of ATPase activity
          positive regulation of cell adhesion
          positive regulation of stress fiber assembly
          actin filament capping
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle contraction
          actin binding
          structural constituent of cytoskeleton
          protein homodimerization activity
          protein N-terminus binding
          actin filament binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164248.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164248.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164249.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164249.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164250.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164250.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164251.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164251.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164255.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_001164255.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_024427.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 NM_024427.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021353.mm.1 TC0900002541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164248.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164248.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164249.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164249.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164250.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164250.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164251.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164251.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164255.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_001164255.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_024427.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 NM_024427.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021352.mm.1 TC0900002541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_001164248.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_001164248.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_001164250.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_001164250.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_001164251.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_001164251.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_001164255.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_001164255.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_024427.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 NM_024427.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021350.mm.1 TC0900002541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164248.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164248.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164249.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164249.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164250.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164250.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164251.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164251.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164255.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_001164255.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_024427.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 NM_024427.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021354.mm.1 TC0900002541.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 684484 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline