Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00612628_m1
          See other TSNAXIP1 GE Assays ›
          Gene Symbol:
          Tsnaxip1
          Gene Name:
          translin-associated factor X (Tsnax) interacting protein 1
          Gene Aliases:
          1700016K08Rik, TXI1
          Chromosome Location:
          Chr.8: 105827711 - 105844676 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00612628_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsnaxip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024445.4 NP_077765.1 1 - 2 83 65
          GenBank mRNA AF343750.1 - 1 - 2 55 65
          AK006041.1 - 1 - 2 84 65
          BC100431.1 - 1 - 2 96 30917958 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001229.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsnaxip1

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          biological_process
          cell differentiation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P195825 AK006041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P195825 NM_024445.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P195825 AF343750.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P195825 AK006041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P195825 NM_024445.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P195825 AF343750.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129894 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149810 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167959 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121086 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P195825 AK006041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P195825 NM_024445.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P195825 AF343750.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P195825 AK006041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P195825 NM_024445.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P195825 AF343750.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26136169 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121086 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06627789 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167959 - -
          Whole Mouse Genome G4122A A_51_P195825 AK006041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195825 NM_024445.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195825 AF343750.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195825 AK006041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195825 NM_024445.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195825 AF343750.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132644 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167959 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121086 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06873954 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06873959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26136289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07373256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07090891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06970106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06802081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07373254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06703145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07450006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07211585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26136204 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574886 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574890 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574881 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574891 - -
          Affymetrix HT MG-430 PM Array Plate 1427018_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574880 - -
          Mouse Genome 430 2.0 Array 1427018_at - -
          Mouse Genome 430 2.0 Array 1427018_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72236_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72236_at - -
          Affymetrix Mouse Clariom S Array PSR0800009785.mm.1 NM_024445.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009785.mm.1 NM_024445.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009785.mm.1 TC0800001229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009786.mm.1 NM_024445.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009786.mm.1 NM_024445.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009786.mm.1 TC0800001229.mm.1 MTA TC
          Mouse Expression Set 430 1427018_PM_at - -
          Mouse Expression Set 430 1427018_at - -
          Mouse Genome 430A 2.0 Array 1427018_at - -
          Mouse Genome 430A 2.0 Array 1427018_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427018_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427018_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 615427 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1770270 - -
          Illumina mouse-6 v1.0 expression beadchip 6180577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770270 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224276 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240576 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224276 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240576 - -
          mouse-6 v1.1 (Illumina) scl000713.1_43-S - -
          mouse-6 v1.1 (Illumina) scl33403.16.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 1770270 - -
          Illumina Sentrix mouse-6.v1 bead array 6180577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000713.1_43-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33403.16.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770270 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1240576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2881498 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2881498 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240576 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline