Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00613143_s1
          See other ZCCHC3 GE Assays ›
          Gene Symbol:
          Zcchc3
          Gene Name:
          zinc finger, CCHC domain containing 3
          Gene Aliases:
          2810406K24Rik
          Chromosome Location:
          Chr.2: 152411956 - 152415044 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00613143_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zcchc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175126.4 NP_780335.1 1 1117 90
          GenBank mRNA AK053859.1 - 1 854 90
          BC055755.1 - 1 1100 6412435 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004899.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zcchc3

          Panther Classification:

          Molecular Function -

          viral or transposable element protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749217 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749216 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749223 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479132 BC055755.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479132 AK053859.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479132 NM_175126.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479132 BC055755.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479132 AK053859.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P479132 NM_175126.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479132 BC055755.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479132 AK053859.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479132 NM_175126.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479132 BC055755.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479132 AK053859.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P479132 NM_175126.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479132 BC055755.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479132 AK053859.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479132 NM_175126.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479132 BC055755.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479132 AK053859.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P479132 NM_175126.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749195 - -
          Whole Mouse Genome G4122A A_51_P479132 BC055755.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479132 AK053859.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479132 NM_175126.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479132 BC055755.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479132 AK053859.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P479132 NM_175126.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106361 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01318125 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01694329 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01694323 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01318127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749217 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749216 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749223 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21749224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01368648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01368651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02124660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02124655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01472786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02078261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01322405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02124656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01368649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10488627 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488629 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488628 - -
          Affymetrix HT MG-430 PM Array Plate 1428402_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428401_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10488626 - -
          Mouse Genome 430 2.0 Array 1428402_at - -
          Mouse Genome 430 2.0 Array 1428402_PM_at - -
          Mouse Genome 430 2.0 Array 1428401_PM_at - -
          Mouse Genome 430 2.0 Array 1428401_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67917_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67917_at - -
          Affymetrix Mouse Clariom S Array PSR0200041615.mm.1 NM_175126.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041615.mm.1 NM_175126.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041615.mm.1 TC0200004899.mm.1 MTA TC
          Mouse Expression Set 430 1428402_at - -
          Mouse Expression Set 430 1428401_PM_at - -
          Mouse Expression Set 430 1428402_PM_at - -
          Mouse Expression Set 430 1428401_at - -
          Murine Genome U74v2 Set 103374_PM_at - -
          Murine Genome U74v2 Set 103375_at - -
          Murine Genome U74v2 Set 103375_PM_at - -
          Murine Genome U74v2 Set 103374_at - -
          Mouse Genome 430A 2.0 Array 1428402_at - -
          Mouse Genome 430A 2.0 Array 1428401_PM_at - -
          Mouse Genome 430A 2.0 Array 1428401_at - -
          Mouse Genome 430A 2.0 Array 1428402_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428402_at - -
          Affymetrix HT Mouse Genome 430A Array 1428401_at - -
          Affymetrix HT Mouse Genome 430A Array 1428402_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428401_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 853542 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4780278 - -
          MouseWG-6 v2 Expression BeadChip 1990035 - -
          Illumina mouse-6 v1.0 expression beadchip 1990035 - -
          Illumina mouse-6 v1.0 expression beadchip 580438 - -
          Illumina mouse-6 v1.0 expression beadchip 4780278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000580438 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990035 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2476904 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2437173 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248465 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2476904 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2437173 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248465 - -
          mouse-6 v1.1 (Illumina) scl0067917.1_135-S - -
          mouse-6 v1.1 (Illumina) scl067917.2_176-S - -
          mouse-6 v1.1 (Illumina) scl067917.1_76-S - -
          Illumina Sentrix mouse-6.v1 bead array 1990035 - -
          Illumina Sentrix mouse-6.v1 bead array 580438 - -
          Illumina Sentrix mouse-6.v1 bead array 4780278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067917.1_76-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067917.1_135-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067917.2_176-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 580438 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990035 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2437173 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2951955 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248465 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248465 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2437173 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2951955 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067917.1_76-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline