Search
Search

Aifm3| Gene Symbol: Aifm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291070.1 | NP_001277999.1 | 8 - 9 | 958 | 84 | |
| NM_175178.4 | NP_780387.2 | 10 - 11 | 1364 | 84 | ||
| GenBank mRNA | AK012958.1 | - | 6 - 7 | 716 | 84 | |
| AK049928.1 | - | 10 - 11 | 1195 | 84 | ||
| AK158809.1 | - | 9 - 10 | 1026 | 84 | ||
| BC046517.1 | - | 10 - 11 | 1350 | 84 | ||
| BC096476.1 | - | 9 - 10 | 1139 | 4221224 | 84 | |
| BC120685.1 | - | 9 - 10 | 993 | 40129608 | 84 | |
Set Membership: |
|