Search
Search

Tasp1| Gene Symbol: Tasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159640.2 | NP_001153112.1 | 9 - 10 | 916 | 63 | |
| NM_001159641.2 | NP_001153113.1 | 9 - 10 | 916 | 63 | ||
| NM_175225.5 | NP_780434.1 | 9 - 10 | 1183 | 63 | ||
| NR_110350.1 | 8 - 9 | 1103 | 63 | |||
| NR_110351.1 | 9 - 10 | 916 | 63 | |||
| NR_110352.1 | 8 - 9 | 847 | 63 | |||
| GenBank mRNA | AK015624.1 | - | 9 - 10 | 916 | 63 | |
| AK036512.1 | - | 9 - 10 | 1139 | 63 | ||
| AK132709.1 | - | 8 - 9 | 833 | 63 | ||
| AK146320.1 | - | 9 - 10 | 899 | 63 | ||
| AK160983.1 | - | 8 - 9 | 1064 | 63 | ||
| BC024597.1 | - | 9 - 10 | 903 | 4036044 | 63 | |
Set Membership: |
|