Search
Search
| Gene Symbol: Prtg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175485.4 | NP_780694.3 | 14 - 15 | 2623 | 78 | |
| GenBank mRNA | AK039115.1 | - | 1 - 2 | 139 | 78 | |
| AK083540.1 | - | 12 - 13 | 2059 | 78 | ||
| AY630257.1 | - | 14 - 15 | 2693 | 78 | ||
| BC116336.1 | - | 12 - 13 | 2056 | 78 | ||
| BC116337.1 | - | 12 - 13 | 2056 | 78 | ||
| DQ360114.1 | - | 14 - 15 | 2623 | 78 | ||
Set Membership: |
|