Search
Search
| Gene Symbol: A2m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175628.3 | NP_783327.2 | 1 - 2 | 223 | 151 | |
| GenBank mRNA | AY185125.1 | - | 1 - 2 | 172 | 151 | |
| BC072642.1 | - | 1 - 2 | 163 | 6839761 | 151 | |
Set Membership: |
|