Search
Search

Esrp2| Gene Symbol: Esrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176838.2 | NP_789808.1 | 8 - 9 | 1166 | 68 | |
| GenBank mRNA | AK088755.1 | - | 8 - 9 | 1036 | 68 | |
| AK168488.1 | - | 8 - 9 | 1014 | 68 | ||
| BC031444.1 | - | 8 - 9 | 1166 | 4222237 | 68 | |
| BC096581.1 | - | 8 - 9 | 1003 | 68 | ||
Set Membership: |
|