Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617298_m1
          See other BC048502 GE Assays ›
          Gene Symbol:
          BC048502
          Gene Name:
          cDNA sequence BC048502
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 103438950 - 103450079 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617298_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC048502
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177631.3 NP_808299.1 5 - 6 503 100
          GenBank mRNA AK131692.1 - 5 - 6 515 100
          BC048502.1 - 5 - 6 503 6740942 100

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BC048502

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P133538 NM_177631.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P133538 AK131692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P133538 BC048502.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P133538 NM_177631.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P133538 AK131692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P133538 BC048502.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741951 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915644 AK081892.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915644 AK153708.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915644 BC020030.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915644 BC026661.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915644 BC044805.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915644 BC095966.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915644 NM_001112744.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915644 XM_149562.8 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915644 XM_902459.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 559200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111152 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121110 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P133538 NM_177631.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P133538 AK131692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P133538 BC048502.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P133538 NM_177631.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P133538 AK131692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P133538 BC048502.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30504176 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08489704 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08489716 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121110 - -
          Whole Mouse Genome G4122A A_52_P133538 NM_177631.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P133538 AK131692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P133538 BC048502.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P133538 NM_177631.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P133538 AK131692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P133538 BC048502.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124805 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134803 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121110 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11973912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11973904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12150969 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12049404 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11973906 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12150968 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12344238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12344236 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11973908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30504089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12056032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11845385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12217041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12158915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12315027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12443202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12443198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12448888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11848262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12056021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12060578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30504120 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10433159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433155 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433158 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433154 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433160 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433157 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433156 - -
          Affymetrix Mouse Gene 1.1 ST Array 10433153 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 618993 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5420338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2729935 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2729935 - -
          mouse-6 v1.1 (Illumina) scl46655.7.1_64-S - -
          Illumina Sentrix mouse-6.v1 bead array 5420338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46655.7.1_64-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2960128 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2960128 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline