Search
Search

Dnajb2| Gene Symbol: Dnajb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159883.1 | NP_001153355.1 | 1 - 2 | 171 | 95 | |
| NM_020266.2 | NP_064662.2 | 1 - 2 | 171 | 95 | ||
| NM_178055.4 | NP_835156.1 | 1 - 2 | 171 | 95 | ||
| GenBank mRNA | AK076061.1 | - | 1 - 2 | 107 | 95 | |
| AK156083.1 | - | 1 - 2 | 140 | 95 | ||
| AK171494.1 | - | 1 - 2 | 117 | 95 | ||
| BY073498.1 | - | 1 - 2 | 174 | 95 | ||
Set Membership: |
|