Search
Search

Arrb1| Gene Symbol: Arrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177231.2 | NP_796205.1 | 7 - 8 | 776 | 136 | |
| NM_178220.3 | NP_835738.1 | 7 - 8 | 776 | 136 | ||
| GenBank mRNA | AK052588.1 | - | 7 - 8 | 553 | 136 | |
| AK090090.1 | - | 7 - 8 | 603 | 136 | ||
| AK144054.1 | - | 7 - 8 | 575 | 136 | ||
| AK147508.1 | - | 7 - 8 | 777 | 136 | ||
| AK147566.1 | - | 7 - 8 | 798 | 136 | ||
| AK165514.1 | - | 7 - 8 | 575 | 136 | ||
| BC108969.1 | - | 7 - 8 | 543 | 40047224 | 136 | |
| BC108970.1 | - | 7 - 8 | 543 | 40047225 | 136 | |
Set Membership: |
|