Search
Search
| Gene Symbol: E2f7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178609.4 | NP_848724.2 | 7 - 8 | 1343 | 88 | |
| GenBank mRNA | AK031078.1 | - | 7 - 8 | 1341 | 88 | |
| AK045040.1 | - | 7 - 8 | 1308 | 88 | ||
| AK143686.1 | - | 7 - 8 | 1316 | 88 | ||
| AY463359.1 | - | 7 - 8 | 1340 | 88 | ||
| BC145429.1 | - | 7 - 8 | 1296 | 9053987 | 88 | |
| BC150772.1 | - | 7 - 8 | 1296 | 9087683 | 88 | |
Set Membership: |
|