Search
Search
| Gene Symbol: Krr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178610.4 | NP_848725.2 | 6 - 7 | 761 | 124 | |
| GenBank mRNA | AK011972.1 | - | 6 - 7 | 671 | 124 | |
| AK012108.1 | - | 6 - 7 | 699 | 124 | ||
| AK030560.1 | - | 6 - 7 | 673 | 124 | ||
| AK032472.1 | - | 6 - 7 | 673 | 124 | ||
| AK036447.1 | - | 5 - 6 | 759 | 124 | ||
| AK041854.1 | - | 6 - 7 | 673 | 124 | ||
| AK146926.1 | - | 6 - 7 | 679 | 124 | ||
| AK168675.1 | - | 6 - 7 | 673 | 124 | ||
| BC089510.1 | - | 6 - 7 | 678 | 30246541 | 124 | |
| BC094236.1 | - | 6 - 7 | 662 | 124 | ||
Set Membership: |
|