| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC137970.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC137971.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC145068.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC079583.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC088988.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC137970.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC137971.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC145068.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC079583.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P238734 |
BC088988.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
233906 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
106487 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
964520 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
515097 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
229988 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
852900 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
57952 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829221 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58928 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
630342 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
181289 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
226729 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
723126 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
583968 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
912267 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760172 |
AF030131.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760172 |
AK045470.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760172 |
AK173185.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760172 |
BC060113.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760172 |
BC060696.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
760172 |
NM_021506.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
135237 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
68848 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
116818 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P128150 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC137970.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC137971.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC145068.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC079583.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC088988.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC137970.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC137971.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC145068.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC079583.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P238734 |
BC088988.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161678 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161668 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161682 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161661 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161659 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161666 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161662 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161676 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161673 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161681 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161677 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161664 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161656 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161679 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28161671 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07506637 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P128150 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07506571 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07506586 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC137970.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC137971.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC145068.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC079583.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC088988.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC137970.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC137971.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC145068.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC079583.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P238734 |
BC088988.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P65703 |
BC137970.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P65703 |
BC137971.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P65703 |
BC145068.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P65703 |
BC079583.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P65703 |
BC088988.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P114948 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P128150 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09483422 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161697 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161732 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161721 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161748 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161664 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161762 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161746 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161691 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161780 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161817 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161735 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161654 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161710 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161686 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161692 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161718 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161731 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161738 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161787 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161602 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161668 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161699 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161798 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161775 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161804 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161808 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161754 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161741 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161751 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161649 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161753 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161799 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161659 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161662 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161726 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161707 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161711 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161651 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161708 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161778 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161706 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161765 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161736 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161656 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161661 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161715 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161648 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161723 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161603 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161760 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161700 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161701 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161688 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161652 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161781 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161761 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161752 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161809 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161785 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161724 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161818 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161704 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161676 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161820 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161678 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161766 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161694 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161666 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161685 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161677 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28161801 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161697 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161692 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09481691 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161664 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09248839 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09207075 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09483423 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161679 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161666 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161668 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161696 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09248838 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161678 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161804 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161659 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161662 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161673 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161694 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09671391 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161676 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09207074 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161686 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161704 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09106273 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161677 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161681 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09285515 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161654 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09483437 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161656 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161691 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161682 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161680 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09248837 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161661 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161702 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28161809 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09481690 |
- |
- |