Search
Search

Radil| Gene Symbol: Radil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289588.1 | NP_001276517.1 | 7 - 8 | 1906 | 105 | |
| NM_178702.4 | NP_848817.2 | 7 - 8 | 1906 | 105 | ||
| GenBank mRNA | AK052964.1 | - | 7 - 8 | 1817 | 105 | |
| AK079539.1 | - | 7 - 8 | 1341 | 105 | ||
| AK083410.1 | - | 7 - 8 | 1924 | 105 | ||
| AK154169.1 | - | 7 - 8 | 1847 | 105 | ||
| AK162917.1 | - | 7 - 8 | 1887 | 105 | ||
| AK173277.1 | - | 7 - 8 | 1870 | 105 | ||
| BC056483.1 | - | 7 - 8 | 1858 | 5696312 | 105 | |
Set Membership: |
|