Search
Search

Obsl1| Gene Symbol: Obsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178884.5 | NP_849215.3 | 3 - 4 | 1766 | 108 | |
| GenBank mRNA | AK029863.1 | - | 3 - 4 | 1556 | 108 | |
| AK031074.1 | - | 3 - 4 | 1767 | 108 | ||
| AK048144.1 | - | 3 - 4 | 958 | 108 | ||
| AK162473.1 | - | 3 - 4 | 1676 | 108 | ||
| BC051643.1 | - | 1 - 2 | 318 | 108 | ||
Set Membership: |
|