Search
Search

Sh2d7| Gene Symbol: Sh2d7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173778.3 | NP_776139.1 | 2 - 3 | 348 | 103 | |
| GenBank mRNA | AK030178.1 | - | 2 - 3 | 349 | 103 | |
| BC116842.1 | - | 2 - 3 | 271 | 40126161 | 103 | |
| BC116844.1 | - | 2 - 3 | 271 | 40126163 | 103 | |
Set Membership: |
|