Search
Search

Ankar| Gene Symbol: Ankar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176980.4 | NP_795954.2 | 3 - 4 | 1180 | 140 | |
| GenBank mRNA | AK030065.1 | - | 3 - 4 | 1181 | 140 | |
| BC132415.1 | - | 3 - 4 | 1155 | 40130692 | 140 | |
| BC145253.1 | - | 3 - 4 | 1155 | 9053805 | 140 | |
Set Membership: |
|