| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500018348.mm.1 |
NM_001199632.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500018348.mm.1 |
NM_001199632.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500018348.mm.1 |
NM_177076.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500018348.mm.1 |
NM_177076.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500018348.mm.1 |
TC0500002018.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500018349.mm.1 |
NM_001199632.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500018349.mm.1 |
NM_001199632.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500018349.mm.1 |
NM_177076.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500018349.mm.1 |
NM_177076.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500018349.mm.1 |
TC0500002018.mm.1 |
MTA TC |