Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00623339_g1
          See other PHF8 GE Assays ›
          Gene Symbol:
          Phf8
          Gene Name:
          PHD finger protein 8
          Gene Aliases:
          9830141C09Rik, mKIAA1111
          Chromosome Location:
          Chr.X: 151520612 - 151633859 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00623339_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phf8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113354.1 NP_001106825.1 3 - 4 1047 87
          NM_177201.5 NP_796175.1 3 - 4 1047 87
          GenBank mRNA AK036609.1 - 3 - 4 421 87
          AK040943.1 - 3 - 4 416 87
          AK040969.1 - 3 - 4 1048 87
          AK122447.1 - 2 - 3 138 87
          AK134137.1 - 3 - 4 384 87
          AK147939.1 - 3 - 4 434 87
          BC138899.1 - 3 - 4 316 8861921 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001520.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Phf8

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          histone H3-K9 demethylation
          histone H4-K20 demethylation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase I promoter
          oxidation-reduction process
          negative regulation of chromatin silencing at rDNA
          histone H3-K36 demethylation
          histone H3-K27 demethylation
          chromatin binding
          iron ion binding
          zinc ion binding
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
          histone demethylase activity
          histone demethylase activity (H3-K9 specific)
          methylated histone binding
          histone demethylase activity (H4-K20 specific)
          histone demethylase activity (H3-K36 specific)
          histone demethylase activity (H3-K27 specific)

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00009507.mm.1 NM_001113354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009507.mm.1 NM_001113354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009507.mm.1 NM_177201.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009507.mm.1 NM_177201.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009507.mm.1 TC0X00001520.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009510.mm.1 NM_001113354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009510.mm.1 NM_001113354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009510.mm.1 NM_177201.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009510.mm.1 NM_177201.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009510.mm.1 TC0X00001520.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009511.mm.1 NM_001113354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009511.mm.1 NM_001113354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009511.mm.1 NM_177201.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009511.mm.1 NM_177201.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009511.mm.1 TC0X00001520.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009502.mm.1 NM_001113354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009502.mm.1 NM_001113354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009502.mm.1 NM_177201.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009502.mm.1 NM_177201.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009502.mm.1 TC0X00001520.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009505.mm.1 NM_001113354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009505.mm.1 NM_001113354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009505.mm.1 NM_177201.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009505.mm.1 NM_177201.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009505.mm.1 TC0X00001520.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009506.mm.1 NM_001113354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009506.mm.1 NM_001113354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009506.mm.1 NM_177201.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009506.mm.1 NM_177201.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009506.mm.1 TC0X00001520.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00009509.mm.1 NM_001113354.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009509.mm.1 NM_001113354.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009509.mm.1 NM_177201.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009509.mm.1 NM_177201.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00009509.mm.1 TC0X00001520.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 762861 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline