Search
Search

Enpp6| Gene Symbol: Enpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177304.3 | NP_796278.1 | 6 - 7 | 1045 | 68 | |
| GenBank mRNA | AK014741.1 | - | 1 - 2 | 101 | 68 | |
| AK038460.1 | - | 6 - 7 | 1017 | 68 | ||
| AK046797.1 | - | 6 - 7 | 1046 | 68 | ||
| AK046881.1 | - | 6 - 7 | 1050 | 68 | ||
| AK079352.1 | - | 5 - 6 | 1387 | 68 | ||
| AK079642.1 | - | 6 - 7 | 1045 | 68 | ||
| BC096376.1 | - | 6 - 7 | 1016 | 4237600 | 68 | |
Set Membership: |
|