Search
Search

Vwa3a| Gene Symbol: Vwa3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177697.3 | NP_808365.2 | 24 - 25 | 2854 | 111 | |
| GenBank mRNA | AK086942.1 | - | 23 - 24 | 2452 | 111 | |
| AK136898.1 | - | 23 - 24 | 2453 | 111 | ||
Set Membership: |
|