Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00625297_m1
          See other BC049635 GE Assays ›
          Gene Symbol:
          BC049635
          Gene Name:
          cDNA sequence BC049635
          Gene Aliases:
          4931430D02
          Chromosome Location:
          Chr.4: 42868003 - 42874838 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00625297_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC049635
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177785.4 NP_808453.1 4 - 5 913 106
          GenBank mRNA AK077002.1 - 4 - 5 914 106
          AK133100.1 - 2 - 3 835 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002510.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BC049635

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964163 BC049635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964163 NM_177785.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964163 AK077002.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964163 AK133100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337388 BC049635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337388 NM_177785.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337388 AK077002.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337388 AK133100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337388 BC049635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337388 NM_177785.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337388 AK077002.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P337388 AK133100.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545298 AK040527.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386137 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64709 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124149 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337388 BC049635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337388 NM_177785.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337388 AK077002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337388 AK133100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337388 BC049635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337388 NM_177785.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337388 AK077002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P337388 AK133100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964163 BC049635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964163 NM_177785.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964163 AK077002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964163 AK133100.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22868680 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124149 - -
          Whole Mouse Genome G4122A A_52_P337388 BC049635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337388 NM_177785.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337388 AK077002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337388 AK133100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337388 BC049635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337388 NM_177785.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337388 AK077002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337388 AK133100.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127249 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124149 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03556188 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03556205 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03531968 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03556190 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03556187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22868595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03305536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03556205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03234755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03556197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03556190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03556191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03479083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03234752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04074745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03251177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03913247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03556192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03305535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22868689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03234754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03523523 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512388 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512385 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512387 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512386 - -
          Affymetrix Mouse Gene 1.1 ST Array 10512384 - -
          Affymetrix Mouse Clariom S Array PSR0400020249.mm.1 TC0400002510.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020249.mm.1 NM_177785.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020249.mm.1 NM_177785.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020251.mm.1 TC0400002510.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020251.mm.1 NM_177785.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020251.mm.1 NM_177785.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020254.mm.1 TC0400002510.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020254.mm.1 NM_177785.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020254.mm.1 NM_177785.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 930109 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 940204 - -
          Illumina mouse-6 v1.0 expression beadchip 940204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000940204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1260363 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260363 - -
          mouse-6 v1.1 (Illumina) scl24413.5.4_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 940204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24413.5.4_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 940204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1260363 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3125842 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1260363 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3125842 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline