Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00625490_m1
          See other LRRC16A GE Assays ›
          Gene Symbol:
          Lrrc16a
          Gene Name:
          leucine rich repeat containing 16A
          Gene Aliases:
          1110037D04Rik, AI425970, CARMIL, D130057M20, Lrrc16
          Chromosome Location:
          Chr.13: 24012481 - 24280790 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00625490_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc16a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026825.3 NP_081101.3 4 - 5 440 104
          GenBank mRNA AK051570.1 - 4 - 5 440 104
          AY437876.1 - 4 - 5 445 104
          BC150819.1 - 4 - 5 366 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001654.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrc16a

          Gene Ontology Categories:

          Function(s) Process(es)

          cell migration
          lamellipodium assembly
          positive regulation of cell migration
          positive regulation of actin filament polymerization
          ruffle organization
          urate metabolic process
          positive regulation of stress fiber assembly
          barbed-end actin filament uncapping
          positive regulation of substrate adhesion-dependent cell spreading
          positive regulation of lamellipodium organization
          negative regulation of barbed-end actin filament capping
          protein complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300010290.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010290.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010290.mm.1 TC1300001654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010297.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010297.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010297.mm.1 TC1300001654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010292.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010292.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010292.mm.1 TC1300001654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010287.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010287.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010287.mm.1 TC1300001654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010296.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010296.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010296.mm.1 TC1300001654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010289.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010289.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010289.mm.1 TC1300001654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010291.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010291.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010291.mm.1 TC1300001654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010294.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010294.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010294.mm.1 TC1300001654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010295.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010295.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010295.mm.1 TC1300001654.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010299.mm.1 NM_026825.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010299.mm.1 NM_026825.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010299.mm.1 TC1300001654.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline