Search
Search
| Gene Symbol: Tnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177839.3 | NP_808507.2 | 20 - 21 | 4509 | 64 | |
| GenBank mRNA | AF455756.1 | - | 20 - 21 | 4476 | 64 | |
| AJ580920.1 | - | 17 - 18 | 3636 | 64 | ||
| AK048033.1 | - | 13 - 14 | 2624 | 64 | ||
| BC138335.1 | - | 20 - 21 | 4470 | 8861355 | 64 | |
| BC138336.1 | - | 20 - 21 | 4470 | 8861356 | 64 | |
Set Membership: |
|