Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00625709_m1
          See other DCDC5 GE Assays ›
          Gene Symbol:
          Dcdc5
          Gene Name:
          doublecortin domain containing 5
          Gene Aliases:
          4732421G10Rik, BC048594, EG436559
          Chromosome Location:
          Chr.2: 106042135 - 106408603 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00625709_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dcdc5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK028639.1 - 2 - 3 742 94
          BC141310.1 - 2 - 3 579 9055922 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001591.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249104 AK028639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P249104 AK028639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031460 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249104 AK028639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P249104 AK028639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2031460 - -
          Whole Mouse Genome G4122A A_51_P249104 AK028639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249104 AK028639.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P01757542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02039462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01515578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01997862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02261585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01799518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01515576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01515580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01239639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01957315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02253992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01475067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01957321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01242599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01757538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01497272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02261582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01730442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02007671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01515583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01957322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01210250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01262473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01336210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01805396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01782179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01799516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02123322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01592764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01975615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01809341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01757547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01799517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01262480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01778150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02253994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01302526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01515568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01592749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02039460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21500098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21499910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02039461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02133500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01679906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01592755 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474358 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474360 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474355 - -
          Affymetrix Mouse Clariom S Array PSR0200011996.mm.1 TC0200001591.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011995.mm.1 TC0200001591.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200011993.mm.1 TC0200001591.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 309588 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline