Search
Search

Stk32a| Gene Symbol: Stk32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178749.3 | NP_848864.1 | 2 - 3 | 291 | 72 | |
| GenBank mRNA | AK036266.1 | - | 2 - 3 | 292 | 72 | |
| AK042599.1 | - | 2 - 3 | 405 | 72 | ||
| AK044474.1 | - | 2 - 3 | 301 | 72 | ||
| BC055002.1 | - | 311 | 6313304 | 72 | ||
Set Membership: |
|