Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00626861_m1
          See other PLD4 GE Assays ›
          Gene Symbol:
          Pld4
          Gene Name:
          phospholipase D family, member 4
          Gene Aliases:
          AI132321, thss
          Chromosome Location:
          Chr.12: 112760655 - 112768988 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00626861_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pld4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178911.4 NP_849242.1 2 - 3 207 68
          GenBank mRNA AK045459.1 - 2 - 3 166 68
          AK079549.1 - 2 - 3 166 68
          AK079878.1 - 2 - 3 209 68
          AK151551.1 - 2 - 3 441 68
          AK152969.1 - 1 - 2 54 68
          AK170362.1 - 2 - 3 164 68
          BC058565.1 - 2 - 3 157 6442038 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001272.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pld4

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          lipid metabolic process
          phagocytosis
          lipid catabolic process
          catalytic activity
          phospholipase D activity
          protein binding
          hydrolase activity
          N-acylphosphatidylethanolamine-specific phospholipase D activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK170362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 BC058565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK152969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK079878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK151551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK170362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 BC058565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK152969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK079878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK151551.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95131 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158352 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK170362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 BC058565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK152969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK079878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK151551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK170362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 BC058565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK152969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK079878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK151551.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158352 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07755088 - -
          Whole Mouse Genome G4122A A_52_P591153 AK170362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591153 BC058565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591153 AK152969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591153 AK079878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591153 AK151551.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591153 AK170362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591153 BC058565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591153 AK152969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591153 AK079878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591153 AK151551.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115714 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158352 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09877961 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10304230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10304240 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09963115 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10304250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09899983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10402905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10304243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09979466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09979460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10402904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10304236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10399537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10399541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10399555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10374915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09979475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09979472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10304246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28752292 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10398913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398909 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398911 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398916 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398918 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398908 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398910 - -
          Affymetrix HT MG-430 PM Array Plate 1433678_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10398907 - -
          Mouse Genome 430 2.0 Array 1433678_at - -
          Mouse Genome 430 2.0 Array 1433678_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 104759_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 104759_at - -
          Affymetrix Mouse Clariom S Array PSR1200009155.mm.1 NM_178911.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009155.mm.1 NM_178911.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009155.mm.1 TC1200001272.mm.1 MTA TC
          Mouse Expression Set 430 1433678_at - -
          Mouse Expression Set 430 1433678_PM_at - -
          Murine Genome U74v2 Set 162043_i_at - -
          Murine Genome U74v2 Set 103299_at - -
          Murine Genome U74v2 Set 162043_PM_i_at - -
          Murine Genome U74v2 Set 103299_PM_at - -
          Murine Genome U74v2 Set 168401_PM_f_at - -
          Murine Genome U74v2 Set 168401_f_at - -
          Mouse Genome 430A 2.0 Array 1433678_PM_at - -
          Mouse Genome 430A 2.0 Array 1433678_at - -
          Affymetrix HT Mouse Genome 430A Array 1433678_at - -
          Affymetrix HT Mouse Genome 430A Array 1433678_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 379453 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6590066 - -
          Illumina mouse-6 v1.0 expression beadchip 6590066 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006590066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234223 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234223 - -
          mouse-6 v1.1 (Illumina) scl42725.12.1_79-S - -
          Illumina Sentrix mouse-6.v1 bead array 6590066 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42725.12.1_79-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234223 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234223 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline