Search
Search

Utp15| Gene Symbol: Utp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178918.3 | NP_849249.1 | 9 - 10 | 1237 | 77 | |
| GenBank mRNA | AK049186.1 | - | 9 - 10 | 1237 | 77 | |
| AK162794.1 | - | 9 - 10 | 1241 | 77 | ||
| AK165546.1 | - | 9 - 10 | 1232 | 77 | ||
| AY151261.1 | - | 9 - 10 | 1178 | 77 | ||
| BC034372.1 | - | 5 - 6 | 594 | 4021387 | 77 | |
| BC065057.1 | - | 9 - 10 | 1237 | 5715800 | 77 | |
Set Membership: |
|