Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026268.3 | NP_080544.1 | 1 - 2 | 863 | 152 | |
GenBank mRNA | AK005062.1 | - | 1 - 2 | 864 | 152 | |
AK009131.1 | - | 1 - 2 | 791 | 152 | ||
AK088468.1 | - | 1 - 2 | 864 | 152 | ||
AK088665.1 | - | 1 - 2 | 865 | 152 | ||
AK151126.1 | - | 1 - 2 | 860 | 152 | ||
AK152773.1 | - | 1 - 2 | 860 | 152 | ||
AK159146.1 | - | 1 - 2 | 764 | 152 | ||
AK159502.1 | - | 1 - 2 | 864 | 152 | ||
AK159900.1 | - | 1 - 2 | 764 | 152 | ||
BC003869.1 | - | 1 - 2 | 445 | 3491528 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |