Search
Search

Ddx51| Gene Symbol: Ddx51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027156.3 | NP_081432.2 | 10 - 11 | 1512 | 83 | |
| GenBank mRNA | AK010015.1 | - | 1 - 2 | 116 | 83 | |
| AK141361.1 | - | 10 - 11 | 1493 | 83 | ||
| AK152599.1 | - | 10 - 11 | 1480 | 83 | ||
| AK211681.1 | - | 3 - 4 | 337 | 83 | ||
| BC023700.1 | - | 7 - 8 | 775 | 83 | ||
| BC060646.1 | - | 10 - 11 | 1489 | 6853826 | 83 | |
| BC069876.1 | - | 3 - 4 | 340 | 83 | ||
Set Membership: |
|