Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653073_m1
          See other ZFP65 GE Assays ›
          Gene Symbol:
          Zfp65
          Gene Name:
          zinc finger protein 65
          Gene Aliases:
          KRAB5, Zfp71-rs1, pz114
          Chromosome Location:
          Chr.13: 67705306 - 67729173 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653073_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145622.2 NP_663597.1 3 - 4 338 124
          GenBank mRNA AK041786.1 - 3 - 4 285 124
          BB621429.1 - 3 - 4 339 124
          BC016248.1 - 2 - 3 247 4527362 124
          U29507.1 - 2 - 3 144 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002259.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp65

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P460398 BC016248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P460398 NM_145622.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P460398 BC016248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P460398 NM_145622.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128630 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P460398 BC016248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P460398 NM_145622.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P460398 BC016248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P460398 NM_145622.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460398 BC016248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460398 NM_145622.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460398 BC016248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P460398 NM_145622.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10667140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10500494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10830539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10596264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10667123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29109588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10667125 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10410371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410372 - -
          Affymetrix Mouse Gene 1.1 ST Array 10410370 - -
          Mouse Genome 430 2.0 Array 1424752_x_at - -
          Mouse Genome 430 2.0 Array 1424752_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR1300014755.mm.1 NM_145622.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014755.mm.1 NM_145622.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014755.mm.1 TC1300002259.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014757.mm.1 NM_145622.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014757.mm.1 NM_145622.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014757.mm.1 TC1300002259.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014756.mm.1 NM_145622.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014756.mm.1 NM_145622.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014756.mm.1 TC1300002259.mm.1 MTA TC
          Mouse Expression Set 430 1424752_PM_x_at - -
          Mouse Expression Set 430 1424752_x_at - -
          Murine Genome U74v2 Set 136275_PM_at - -
          Murine Genome U74v2 Set 136275_at - -
          Mouse Genome 430A 2.0 Array 1424752_x_at - -
          Mouse Genome 430A 2.0 Array 1424752_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1424752_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1424752_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 638465 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3800100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2511000 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2511000 - -
          mouse-6 v1.1 (Illumina) scl00235907.1_67-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00235907.1_67-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2511000 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2511000 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline