Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00658129_gH
          See other POU5F1 GE Assays ›
          Gene Symbol:
          Pou5f1
          Gene Name:
          POU domain, class 5, transcription factor 1
          Gene Aliases:
          NF-A3, Oct-3, Oct-3/4, Oct-4, Oct3, Oct3/4, Oct4, Otf-3, Otf-4, Otf3, Otf3-rs7, Otf3g, Otf4
          Chromosome Location:
          Chr.17: 35506032 - 35510777 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00658129_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pou5f1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252452.1 NP_001239381.1 2 - 3 208 148
          NM_013633.3 NP_038661.2 2 - 3 570 148
          GenBank mRNA AB221654.1 - 2 - 3 502 148
          AB375269.1 - 2 - 3 147 148
          AB375270.1 - 2 - 3 208 148
          AB375271.1 - 1 - 2 400 148
          AB375272.1 - 2 - 3 174 148
          AB375273.1 - 2 - 3 189 148
          AB375276.1 - 2 - 3 704 148
          AB375277.1 - 1 - 2 166 148
          AK145321.1 - 2 - 3 541 148
          BC068268.1 - 2 - 3 546 30019896 148
          M34381.1 - 2 - 3 551 148
          X52437.1 - 2 - 3 449 148

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Mouse Stem Cell Pluripotency 1700 Inventoried Mouse_Stem_Cell Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pou5f1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          mesodermal cell fate commitment
          endodermal cell fate commitment
          ectodermal cell fate commitment
          endodermal cell fate specification
          trophectodermal cell differentiation
          blastocyst growth
          BMP signaling pathway involved in heart induction
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          multicellular organism development
          response to organic substance
          regulation of gene expression
          stem cell population maintenance
          germ-line stem cell population maintenance
          response to retinoic acid
          somatic stem cell population maintenance
          positive regulation of catenin import into nucleus
          mRNA transcription from RNA polymerase II promoter
          cell fate commitment
          negative regulation of cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of calcium ion-dependent exocytosis
          stem cell differentiation
          positive regulation of protein kinase B signaling
          negative regulation of protein kinase B signaling
          positive regulation of SMAD protein import into nucleus
          cell fate commitment involved in formation of primary germ layer
          cardiac cell fate determination
          negative regulation of gene silencing by miRNA
          regulation of heart induction by regulation of canonical Wnt signaling pathway
          regulation of methylation-dependent chromatin silencing
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription coactivator activity
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          transcription factor binding
          chromatin DNA binding
          ubiquitin protein ligase binding
          miRNA binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          poly(A) RNA binding
          protein heterodimerization activity
          POU domain binding
          HMG box domain binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700007085.mm.1 NM_001252452.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007085.mm.1 NM_001252452.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007085.mm.1 NM_013633.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007085.mm.1 NM_013633.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007085.mm.1 TC1700000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007090.mm.1 NM_001252452.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007090.mm.1 NM_001252452.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007090.mm.1 NM_013633.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007090.mm.1 NM_013633.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007090.mm.1 TC1700000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007088.mm.1 NM_001252452.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007088.mm.1 NM_001252452.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007088.mm.1 NM_013633.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007088.mm.1 NM_013633.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007088.mm.1 TC1700000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007075.mm.1 NM_013633.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007075.mm.1 NM_013633.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007075.mm.1 TC1700000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007082.mm.1 NM_001252452.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007082.mm.1 NM_001252452.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007082.mm.1 TC1700000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007087.mm.1 NM_001252452.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007087.mm.1 NM_001252452.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007087.mm.1 NM_013633.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007087.mm.1 NM_013633.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007087.mm.1 TC1700000691.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007076.mm.1 NM_013633.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007076.mm.1 NM_013633.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007076.mm.1 TC1700000691.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 615439 - -

          Back To Top
           

          Citations (1)

          Tcf3 is an integral component of the core regulatory circuitry of embryonic stem cells.
          Authors
          Cole MF, Johnstone SE, Newman JJ, Kagey MH, Young RA
          Journal
          Genes Dev - Page No 746 (Sat Mar 15 00:00:00 EDT 2008)
          Volume (Year)
          22 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline