Search
Search

Prkar1a| Gene Symbol: Prkar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021880.2 | NP_068680.1 | 3 - 4 | 472 | 91 | |
| GenBank mRNA | AK005039.1 | - | 3 - 4 | 474 | 91 | |
| AK019147.1 | - | 1 - 2 | 107 | 91 | ||
| AK027916.1 | - | 3 - 4 | 439 | 91 | ||
| AK049832.1 | - | 3 - 4 | 465 | 91 | ||
| AK051068.1 | - | 3 - 4 | 473 | 91 | ||
| AK053918.1 | - | 3 - 4 | 473 | 91 | ||
| AK083383.1 | - | 3 - 4 | 453 | 91 | ||
| AK145797.1 | - | 3 - 4 | 473 | 91 | ||
| AK145860.1 | - | 3 - 4 | 440 | 91 | ||
| AK146380.1 | - | 3 - 4 | 459 | 91 | ||
| AK147188.1 | - | 3 - 4 | 464 | 91 | ||
| AK150427.1 | - | 3 - 4 | 465 | 91 | ||
| AK150927.1 | - | 1 - 2 | 105 | 91 | ||
| AK151040.1 | - | 1 - 2 | 105 | 91 | ||
| AK151124.1 | - | 3 - 4 | 431 | 91 | ||
| AK151556.1 | - | 1 - 2 | 105 | 91 | ||
| AK152044.1 | - | 1 - 2 | 105 | 91 | ||
| AK153227.1 | - | 3 - 4 | 431 | 91 | ||
| AK158562.1 | - | 3 - 4 | 810 | 91 | ||
| BC003461.1 | - | 3 - 4 | 443 | 2648357 | 91 | |
| BC005697.1 | - | 3 - 4 | 414 | 3597765 | 91 | |
| HQ412666.1 | - | 4 - 5 | 655 | 91 | ||
| HQ412667.1 | - | 3 - 4 | 471 | 91 | ||
| HQ412668.1 | - | 3 - 4 | 531 | 91 | ||
Set Membership: |
|