Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00660559_m1
          See other RCN3 GE Assays ›
          Gene Symbol:
          Rcn3
          Gene Name:
          reticulocalbin 3, EF-hand calcium binding domain
          Gene Aliases:
          6030455P07Rik, D530026G20Rik, D7Ertd671e, RLP49
          Chromosome Location:
          Chr.7: 45082914 - 45092257 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00660559_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rcn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026555.2 NP_080831.2 5 - 6 807 140
          GenBank mRNA AK003918.1 - 4 - 5 477 140
          AK077943.1 - 5 - 6 747 140
          AK082762.1 - 5 - 6 737 140
          AK086705.1 - 5 - 6 747 141
          BC005487.1 - 4 - 5 644 140
          BC025602.1 - 5 - 6 758 5250505 140
          BC055903.1 - 5 - 6 754 4010685 140
          BC092069.1 - 5 - 6 705 4215821 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002988.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rcn3

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          calcium ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046393 BC025602.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046393 BC055903.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046393 BC005487.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046393 BC092069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046393 AK082762.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939999 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636145 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125724 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046393 BC025602.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046393 BC055903.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046393 BC005487.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046393 BC092069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046393 AK082762.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25091104 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125724 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06181874 - -
          Whole Mouse Genome G4122A A_51_P398683 BC025602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398683 BC005487.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398683 BC092069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398683 AK082762.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398683 AV129283.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P223618 BC025602.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P223618 BC055903.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P223618 BC005487.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P223618 BC092069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P223618 AK082762.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116325 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136172 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125724 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06340080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06192953 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06340077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06605018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25091107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06214168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06027048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05972931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06102693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06027049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06605018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06102691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05972927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05907131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06257038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25091064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563078 - -
          Affymetrix HT MG-430 PM Array Plate 1424382_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563077 - -
          Mouse Genome 430 2.0 Array 1424382_PM_at - -
          Mouse Genome 430 2.0 Array 1424382_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52377_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52377_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700026833.mm.1 NM_026555.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026833.mm.1 NM_026555.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026833.mm.1 TC0700002988.mm.1 MTA TC
          Mouse Expression Set 430 1424382_at - -
          Mouse Expression Set 430 1424382_PM_at - -
          Murine Genome U74v2 Set 96347_PM_at - -
          Murine Genome U74v2 Set 96347_at - -
          Mouse Genome 430A 2.0 Array 1424382_PM_at - -
          Mouse Genome 430A 2.0 Array 1424382_at - -
          Affymetrix HT Mouse Genome 430A Array 1424382_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424382_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 537147 - -
          ABI Mouse Genome Survey Microarray 731197 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5420400 - -
          Illumina mouse-6 v1.0 expression beadchip 5420400 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2723718 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246312 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2723718 - -
          mouse-6 v1.1 (Illumina) scl31391.7.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 5420400 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31391.7.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420400 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2723718 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2723718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline