Search
Search

Lrrc16a| Gene Symbol: Lrrc16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026825.3 | NP_081101.3 | 35 - 36 | 3826 | 62 | |
| GenBank mRNA | AK043591.1 | - | 21 - 22 | 2424 | 62 | |
| AY437876.1 | - | 35 - 36 | 3831 | 62 | ||
| BC012229.1 | - | 14 - 15 | 1855 | 62 | ||
| BC150819.1 | - | 34 - 35 | 3740 | 62 | ||
Set Membership: |
|